Assay Details
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011891.1 | NP_001011891.1 | 6 - 7 | 698 | 163 | |
XM_008767558.1 | XP_008765780.1 | 6 - 7 | 711 | 163 | ||
GenBank mRNA | BC079386.1 | - | 6 - 7 | 698 | 163 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
