Assay Details
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750790.3 | XP_003750838.1 | 4 - 5 | 479 | 79 | |
XM_006246064.2 | XP_006246126.1 | 4 - 5 | 479 | 79 | ||
XM_008767617.1 | XP_008765839.1 | 2 - 3 | 487 | 79 | ||
GenBank mRNA | BC076383.1 | - | 3 - 4 | 440 | 79 | |
BC161824.1 | - | 2 - 3 | 639 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
