Assay Details
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750790.3 | XP_003750838.1 | 2 - 3 | 225 | 135 | |
XM_006246064.2 | XP_006246126.1 | 2 - 3 | 225 | 135 | ||
GenBank mRNA | BC076383.1 | - | 1 - 2 | 121 | 135 | |
BC161824.1 | - | 1 - 2 | 125 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC076383.1, XM_003750790.3, XM_006246064.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
