Assay Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247542.2 | XP_006247604.1 | 10 - 11 | 2336 | 91 | |
GenBank mRNA | EU012436.1 | - | 11 - 12 | 1062 | 91 | |
EU012437.1 | - | 10 - 11 | 911 | 91 | ||
EU012438.1 | - | 11 - 12 | 1130 | 91 | ||
L24096.1 | - | 11 - 12 | 1143 | 91 | ||
L25438.1 | - | 11 - 12 | 1065 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
