Assay Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247542.2 | XP_006247604.1 | 11 - 12 | 2378 | 71 | |
GenBank mRNA | EU012436.1 | - | 12 - 13 | 1104 | 71 | |
EU012437.1 | - | 11 - 12 | 953 | 71 | ||
EU012438.1 | - | 12 - 13 | 1172 | 71 | ||
L24096.1 | - | 12 - 13 | 1185 | 71 | ||
L25438.1 | - | 12 - 13 | 1107 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
G protein-coupled receptor signaling pathway
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
adenylate cyclase-activating G protein-coupled receptor signaling pathway
phospholipase C-activating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
memory
feeding behavior
visual learning
hormone-mediated signaling pathway
regulation of adenylate cyclase activity involved in G protein-coupled receptor signaling pathway
hypothalamus development
adrenal gland development
epithelial cell differentiation
negative regulation of epinephrine secretion
locomotory exploration behavior
response to immobilization stress
fear response
positive regulation of mast cell degranulation
behavioral response to cocaine
behavioral response to ethanol
regulation of synaptic plasticity
behavioral response to pain
corticotropin secretion
response to electrical stimulus
general adaptation syndrome, behavioral process
long-term synaptic potentiation
cellular response to corticotropin-releasing hormone stimulus
negative regulation of neuron death
negative regulation of voltage-gated calcium channel activity
negative regulation of feeding behavior
regulation of corticosterone secretion
G-protein alpha-subunit binding
G protein-coupled receptor activity
protein binding
corticotrophin-releasing factor receptor activity
peptide binding
corticotropin-releasing hormone receptor activity
protein-containing complex binding
corticotropin-releasing hormone binding
response to hypoxia
G protein-coupled receptor signaling pathway
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
adenylate cyclase-activating G protein-coupled receptor signaling pathway
phospholipase C-activating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
memory
feeding behavior
visual learning
hormone-mediated signaling pathway
regulation of adenylate cyclase activity involved in G protein-coupled receptor signaling pathway
hypothalamus development
adrenal gland development
epithelial cell differentiation
negative regulation of epinephrine secretion
locomotory exploration behavior
response to immobilization stress
fear response
positive regulation of mast cell degranulation
behavioral response to cocaine
behavioral response to ethanol
regulation of synaptic plasticity
behavioral response to pain
corticotropin secretion
response to electrical stimulus
general adaptation syndrome, behavioral process
long-term synaptic potentiation
cellular response to corticotropin-releasing hormone stimulus
negative regulation of neuron death
negative regulation of voltage-gated calcium channel activity
negative regulation of feeding behavior
regulation of corticosterone secretion
G-protein alpha-subunit binding
G protein-coupled receptor activity
protein binding
corticotrophin-releasing factor receptor activity
peptide binding
corticotropin-releasing hormone receptor activity
protein-containing complex binding
corticotropin-releasing hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162934 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012770 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012770 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012770 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P688027 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P456197 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63313 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18463 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55918 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10738734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738729 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738738 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10738724 | - | - |
Rat Genome 230 2.0 Array | 1398289_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1387683_at | - | - |
Rat Genome 230 2.0 Array | 1387683_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398289_a_at | - | - |
Affymetrix HT Rat Focus Array | 1398289_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387683_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398289_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387683_at | - | - |
Rat Expression Set 230 | 1398289_PM_a_at | - | - |
Rat Expression Set 230 | 1387683_at | - | - |
Rat Expression Set 230 | 1387683_PM_at | - | - |
Rat Expression Set 230 | 1398289_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10738724 | - | - |
Rat Genome U34 Set | U53486mRNA_s_at | - | - |
Rat Genome U34 Set | U53500_s_at | - | - |
Rat Genome U34 Set | AF039203UTR#1_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21126865 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371683 | - | - |
RatRef-12 Expression BeadChip | ILMN_60679 | - | - |
