Assay Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247542.2 | XP_006247604.1 | 2 - 3 | 1511 | 144 | |
GenBank mRNA | EU012437.1 | - | 3 - 4 | 240 | 144 | |
EU012438.1 | - | 3 - 4 | 240 | 144 | ||
L24096.1 | - | 3 - 4 | 318 | 144 | ||
L25438.1 | - | 3 - 4 | 240 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
