Assay Details
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053474.1 | NP_445926.1 | 9 - 10 | 2807 | 71 | |
GenBank mRNA | AF016252.1 | - | 9 - 10 | 2807 | 71 | |
BC160878.1 | - | 9 - 10 | 2489 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
