Assay Details
Gene Symbol: Doc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031142.1 | NP_112404.1 | 1 - 2 | 529 | 68 | |
XM_008768117.1 | XP_008766339.1 | 1 - 2 | 590 | 68 | ||
GenBank mRNA | U70778.2 | - | 1 - 2 | 529 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
