Assay Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031029.1 | NP_112291.1 | 4 - 5 | 502 | 135 | |
XM_008767594.1 | XP_008765816.1 | 5 - 6 | 1088 | 135 | ||
XM_008767595.1 | XP_008765817.1 | 5 - 6 | 1094 | 135 | ||
GenBank mRNA | FQ222085.1 | - | 5 - 6 | 669 | 135 | |
U95368.1 | - | 4 - 5 | 502 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
