Assay Details
Gene Symbol: Thra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017960.1 | NP_001017960.1 | 2 - 3 | 488 | 105 | |
NM_031134.2 | NP_112396.2 | 1 - 2 | 356 | 105 | ||
GenBank mRNA | CB580585.1 | - | 1 - 2 | 288 | 105 | |
M18028.1 | - | 2 - 3 | 378 | 105 | ||
M31174.1 | - | 570 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
