Assay Details
Gene Symbol: Thra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017960.1 | NP_001017960.1 | 7 - 8 | 1161 | 76 | |
NM_031134.2 | NP_112396.2 | 6 - 7 | 1029 | 76 | ||
GenBank mRNA | M18028.1 | - | 7 - 8 | 1051 | 76 | |
M31174.1 | - | 1243 | 76 | |||
M31177.1 | - | 2 - 3 | 370 | 76 | ||
X07409.1 | - | 5 - 6 | 690 | 76 | ||
X12744.1 | - | 6 - 7 | 693 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
