Assay Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013034.4 | NP_037166.2 | 1 - 2 | 451 | 108 | |
XM_008767950.1 | XP_008766172.1 | 2 - 3 | 507 | 108 | ||
GenBank mRNA | M79450.1 | - | 3 - 4 | 575 | 108 | |
X63253.1 | - | 1 - 2 | 459 | 108 | ||
X63995.1 | - | 2 - 3 | 504 | 108 | ||
Y11024.1 | - | 1 - 2 | 344 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
