Assay Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013034.4 | NP_037166.2 | 3 - 4 | 811 | 89 | |
XM_008767950.1 | XP_008766172.1 | 4 - 5 | 867 | 89 | ||
GenBank mRNA | M79450.1 | - | 5 - 6 | 935 | 89 | |
X63253.1 | - | 3 - 4 | 819 | 89 | ||
X63995.1 | - | 4 - 5 | 864 | 89 | ||
Y11024.1 | - | 3 - 4 | 704 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
