Assay Details
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012013.1 | NP_001012013.1 | 1 - 2 | 252 | 56 | |
XM_006247508.2 | XP_006247570.1 | 2 - 3 | 240 | 56 | ||
XM_006247509.2 | XP_006247571.1 | 2 - 3 | 242 | 56 | ||
XM_006247510.2 | XP_006247572.1 | 2 - 3 | 241 | 56 | ||
XM_006247511.2 | XP_006247573.1 | 2 - 3 | 240 | 56 | ||
XM_006247512.2 | XP_006247574.1 | 2 - 3 | 241 | 56 | ||
XM_008768284.1 | XP_008766506.1 | 3 - 4 | 343 | 56 | ||
XM_008768285.1 | XP_008766507.1 | 2 - 3 | 231 | 56 | ||
XM_008768286.1 | XP_008766508.1 | 2 - 3 | 242 | 56 | ||
XM_008768287.1 | XP_008766509.1 | 2 - 3 | 241 | 56 | ||
GenBank mRNA | BC076387.1 | - | 1 - 2 | 252 | 7109482 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC076387.1, NM_001012013.1, XM_006247508.2, XM_006247509.2, XM_006247510.2, XM_006247511.2, XM_006247512.2, XM_008768284.1, XM_008768285.1, XM_008768286.1, XM_008768287.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
