Assay Details
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105766.1 | NP_001099236.1 | 1 - 2 | 224 | 93 | |
GenBank mRNA | BC169008.1 | - | 1 - 2 | 130 | 93 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC169008.1, NM_001105766.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
