Assay Details
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105766.1 | NP_001099236.1 | 4 - 5 | 1305 | 168 | |
XM_006245868.2 | XP_006245930.1 | 4 - 5 | 851 | 168 | ||
GenBank mRNA | BC169008.1 | - | 4 - 5 | 1211 | 168 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
