Assay Details
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105766.1 | NP_001099236.1 | 7 - 8 | 1716 | 139 | |
XM_006245868.2 | XP_006245930.1 | 7 - 8 | 1262 | 139 | ||
GenBank mRNA | BC169008.1 | - | 6 - 7 | 1809 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
