Assay Details
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172021.2 | NP_742018.2 | 6 - 7 | 938 | 99 | |
XM_006247266.2 | XP_006247328.1 | 5 - 6 | 2085 | 99 | ||
XM_006247267.2 | XP_006247329.1 | 3 - 4 | 1226 | 99 | ||
XM_008767953.1 | XP_008766175.1 | 5 - 6 | 2088 | 99 | ||
GenBank mRNA | AJ278800.1 | - | 5 - 6 | 918 | 99 | |
BC076503.1 | - | 6 - 7 | 938 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
