Assay Details
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172021.2 | NP_742018.2 | 9 - 10 | 1824 | 102 | |
XM_006247266.2 | XP_006247328.1 | 8 - 9 | 2971 | 102 | ||
XM_006247267.2 | XP_006247329.1 | 6 - 7 | 2112 | 102 | ||
XM_008767953.1 | XP_008766175.1 | 8 - 9 | 2974 | 102 | ||
GenBank mRNA | AJ278800.1 | - | 8 - 9 | 1810 | 102 | |
BC076503.1 | - | 9 - 10 | 1824 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
