Assay Details
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270544.1 | NP_001257473.1 | 11 - 12 | 1393 | 104 | |
NM_017322.2 | NP_059018.1 | 11 - 12 | 1393 | 104 | ||
XM_006246317.1 | XP_006246379.1 | 11 - 12 | 1389 | 104 | ||
XM_006246318.2 | XP_006246380.1 | 11 - 12 | 1341 | 104 | ||
GenBank mRNA | L27111.1 | - | 10 - 11 | 1149 | 104 | |
L27112.1 | - | 10 - 11 | 1149 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
release of cytochrome c from mitochondria
positive regulation of protein phosphorylation
protein phosphorylation
protein targeting to mitochondrion
activation of cysteine-type endopeptidase activity involved in apoptotic process
JNK cascade
JUN phosphorylation
central nervous system development
response to xenobiotic stimulus
response to mechanical stimulus
response to toxic substance
response to organic substance
positive regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cell morphogenesis involved in differentiation
response to amine
neuron projection development
positive regulation of prostaglandin biosynthetic process
regulation of protein ubiquitination
positive regulation of prostaglandin secretion
positive regulation of chemokine production
cellular response to UV
regulation of circadian rhythm
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of nitric oxide biosynthetic process
positive regulation of transcription, DNA-templated
regulation of JNK cascade
response to cadmium ion
rhythmic process
positive regulation of nitric-oxide synthase biosynthetic process
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
positive regulation of apoptotic signaling pathway
JUN kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
cysteine-type endopeptidase activator activity involved in apoptotic process
mitogen-activated protein kinase kinase kinase binding
MAPK cascade
release of cytochrome c from mitochondria
positive regulation of protein phosphorylation
protein phosphorylation
protein targeting to mitochondrion
activation of cysteine-type endopeptidase activity involved in apoptotic process
JNK cascade
JUN phosphorylation
central nervous system development
response to xenobiotic stimulus
response to mechanical stimulus
response to toxic substance
response to organic substance
positive regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cell morphogenesis involved in differentiation
response to amine
neuron projection development
positive regulation of prostaglandin biosynthetic process
regulation of protein ubiquitination
positive regulation of prostaglandin secretion
positive regulation of chemokine production
cellular response to UV
regulation of circadian rhythm
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of nitric oxide biosynthetic process
positive regulation of transcription, DNA-templated
regulation of JNK cascade
response to cadmium ion
rhythmic process
positive regulation of nitric-oxide synthase biosynthetic process
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
positive regulation of apoptotic signaling pathway
JUN kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
cysteine-type endopeptidase activator activity involved in apoptotic process
mitogen-activated protein kinase kinase kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P011944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99850 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480222 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P480222 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480222 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P451014 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P217423 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700510453 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78549 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50717 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69074 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85983 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56749 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700510453 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10733166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733170 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733177 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733163 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733171 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733173 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733174 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733172 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733176 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733175 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10733160 | - | - |
Rat Genome 230 2.0 Array | 1380024_at | - | - |
Rat Genome 230 2.0 Array | 1380024_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378237_at | - | - |
Rat Genome 230 2.0 Array | 1368646_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368646_at | - | - |
Rat Genome 230 2.0 Array | 1378237_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368646_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368646_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000040022.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040023.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040024.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040019.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040020.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040018.rn.1 | TC1000004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000040025.rn.1 | TC1000004343.rn.1 | RTA TC |
Rat Expression Set 230 | 1368646_PM_at | - | - |
Rat Expression Set 230 | 1368646_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733160 | - | - |
Rat Genome U34 Set | rc_AI231354_at | - | - |
Rat Genome U34 Set | rc_AI231354_g_at | - | - |
Rat Genome U34 Set | L27112_s_at | - | - |
Rat Genome U34 Set | rc_AI231354_g_at | - | - |
Rat Genome U34 Set | rc_AI231354_at | - | - |
Rat Genome U34 Set | L27112_s_at | - | - |
Rat Genome U34 Set | rc_AI231354_at | - | - |
Rat Genome U34 Set | L27112_s_at | - | - |
Rat Genome U34 Set | rc_AI231354_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21134557 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357949 | - | - |
RatRef-12 Expression BeadChip | ILMN_65467 | - | - |
