Assay Details
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270544.1 | NP_001257473.1 | 8 - 9 | 1135 | 155 | |
NM_001270545.1 | NP_001257474.1 | 8 - 9 | 1135 | 155 | ||
NM_017322.2 | NP_059018.1 | 8 - 9 | 1135 | 155 | ||
XM_006246317.1 | XP_006246379.1 | 8 - 9 | 1131 | 155 | ||
XM_006246318.2 | XP_006246380.1 | 8 - 9 | 1083 | 155 | ||
GenBank mRNA | BC061870.1 | - | 8 - 9 | 1102 | 155 | |
L27111.1 | - | 7 - 8 | 891 | 155 | ||
L27112.1 | - | 7 - 8 | 891 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
