Assay Details
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270544.1 | NP_001257473.1 | 1 - 2 | 210 | 98 | |
NM_001270545.1 | NP_001257474.1 | 1 - 2 | 210 | 98 | ||
NM_017322.2 | NP_059018.1 | 1 - 2 | 210 | 98 | ||
XM_006246317.1 | XP_006246379.1 | 1 - 2 | 206 | 98 | ||
GenBank mRNA | BC061870.1 | - | 1 - 2 | 177 | 98 | |
DN948189.1 | - | 1 - 2 | 210 | 98 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061870.1, NM_001270544.1, NM_001270545.1, NM_017322.2, XM_006246317.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
