Assay Details
Gene Symbol: Polg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107060.1 | NP_001100530.1 | 4 - 5 | 1279 | 93 | |
GenBank mRNA | BC091245.1 | - | 4 - 5 | 800 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
