Assay Details
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012949.2 | NP_037081.2 | 8 - 9 | 923 | 81 | |
XM_006246599.2 | XP_006246661.1 | 8 - 9 | 994 | 81 | ||
XM_006246600.2 | XP_006246662.1 | 8 - 9 | 974 | 81 | ||
GenBank mRNA | BC083566.1 | - | 8 - 9 | 910 | 7193009 | 81 |
FQ214984.1 | - | 8 - 9 | 935 | 81 | ||
FQ215197.1 | - | 4 - 5 | 600 | 81 | ||
FQ215221.1 | - | 7 - 8 | 835 | 81 | ||
FQ215310.1 | - | 4 - 5 | 601 | 81 | ||
FQ215357.1 | - | 8 - 9 | 946 | 81 | ||
FQ215526.1 | - | 8 - 9 | 934 | 81 | ||
FQ215648.1 | - | 8 - 9 | 933 | 81 | ||
FQ215759.1 | - | 8 - 9 | 945 | 81 | ||
FQ215780.1 | - | 4 - 5 | 601 | 81 | ||
FQ215843.1 | - | 7 - 8 | 901 | 81 | ||
FQ215865.1 | - | 8 - 9 | 933 | 81 | ||
FQ216020.1 | - | 4 - 5 | 600 | 81 | ||
FQ216150.1 | - | 7 - 8 | 902 | 81 | ||
FQ216271.1 | - | 4 - 5 | 601 | 81 | ||
FQ216382.1 | - | 8 - 9 | 936 | 81 | ||
FQ216386.1 | - | 7 - 8 | 911 | 81 | ||
FQ216660.1 | - | 7 - 8 | 901 | 81 | ||
FQ216781.1 | - | 4 - 5 | 600 | 81 | ||
FQ217095.1 | - | 8 - 9 | 933 | 81 | ||
FQ223831.1 | - | 4 - 5 | 600 | 81 | ||
FQ224938.1 | - | 8 - 9 | 933 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
