Assay Details
Gene Symbol: Dcaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107057.3 | NP_001100527.1 | 2 - 3 | 527 | 121 | |
GenBank mRNA | BC167045.1 | - | 2 - 3 | 527 | 7112910 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
