Assay Details
Gene Symbol: Trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039378.1 | NP_001034467.1 | 2 - 3 | 138 | 88 | |
GenBank mRNA | AY903234.1 | - | 2 - 3 | 138 | 88 | |
AY903235.1 | - | 2 - 3 | 138 | 88 | ||
AY903236.1 | - | 2 - 3 | 138 | 88 | ||
AY903237.1 | - | 2 - 3 | 138 | 88 | ||
AY903238.1 | - | 2 - 3 | 138 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
