Assay Details
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012923.2 | NP_037055.1 | 1 - 2 | 225 | 103 | |
GenBank mRNA | BC081852.1 | - | 1 - 2 | 196 | 7105829 | 103 |
FQ211577.1 | - | 1 - 2 | 277 | 103 | ||
FQ211975.1 | - | 1 - 2 | 277 | 103 | ||
FQ224502.1 | - | 1 - 2 | 186 | 103 | ||
X70871.1 | - | 1 - 2 | 195 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC081852.1, NM_012923.2, X70871.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
