Assay Details
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012923.2 | NP_037055.1 | 4 - 5 | 832 | 73 | |
GenBank mRNA | BC081852.1 | - | 4 - 5 | 803 | 7105829 | 73 |
FQ211975.1 | - | 4 - 5 | 884 | 73 | ||
FQ224502.1 | - | 4 - 5 | 793 | 73 | ||
X70871.1 | - | 4 - 5 | 802 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
