Assay Details
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012923.2 | NP_037055.1 | 5 - 6 | 921 | 94 | |
GenBank mRNA | BC081852.1 | - | 5 - 6 | 892 | 7105829 | 94 |
FQ211577.1 | - | 4 - 5 | 894 | 94 | ||
FQ211975.1 | - | 5 - 6 | 973 | 94 | ||
FQ224502.1 | - | 5 - 6 | 882 | 94 | ||
X70871.1 | - | 5 - 6 | 891 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
