Assay Details
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246172.2 | XP_006246234.2 | 1 - 2 | 95 | 92 | |
XM_006246173.2 | XP_006246235.2 | 1 - 2 | 95 | 92 | ||
XM_008767718.1 | XP_008765940.1 | 1 - 2 | 159 | 92 | ||
XM_008767720.1 | XP_008765942.1 | 1 - 2 | 158 | 92 | ||
XM_008767721.1 | XP_008765943.1 | 1 - 2 | 158 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
