Assay Details
Gene Symbol: G6pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013098.2 | NP_037230.2 | 2 - 3 | 401 | 116 | |
GenBank mRNA | BC090067.1 | - | 2 - 3 | 401 | 116 | |
D78592.1 | - | 2 - 3 | 413 | 116 | ||
L37333.1 | - | 2 - 3 | 365 | 116 | ||
U07993.1 | - | 2 - 3 | 363 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
