Assay Details
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171996.3 | NP_741993.1 | 1 - 2 | 170 | 76 | |
XM_006246024.2 | XP_006246086.1 | 2 - 3 | 370 | 76 | ||
GenBank mRNA | AF044574.1 | - | 1 - 2 | 92 | 76 | |
BC070959.1 | - | 1 - 2 | 171 | 7097883 | 76 | |
FQ218511.1 | - | 1 - 2 | 145 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
