Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013133.1 | NP_037265.1 | 1 - 2 | 239 | 74 | |
XM_006246378.2 | XP_006246440.1 | 1 - 2 | 391 | 74 | ||
GenBank mRNA | AJ310834.1 | - | 1 - 2 | 55 | 74 | |
AJ310835.1 | - | 1 - 2 | 55 | 74 | ||
AY827463.1 | - | 1 - 2 | 55 | 74 | ||
D00833.1 | - | 1 - 2 | 239 | 74 | ||
X55246.1 | - | 1 - 2 | 143 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
