Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013133.1 | NP_037265.1 | 3 - 4 | 439 | 129 | |
XM_006246378.2 | XP_006246440.1 | 3 - 4 | 591 | 129 | ||
GenBank mRNA | AJ310834.1 | - | 3 - 4 | 255 | 129 | |
AJ310835.1 | - | 3 - 4 | 255 | 129 | ||
AJ310836.1 | - | 2 - 3 | 127 | 129 | ||
AY827463.1 | - | 3 - 4 | 255 | 129 | ||
D00833.1 | - | 3 - 4 | 439 | 129 | ||
X55246.1 | - | 3 - 4 | 343 | 129 | ||
Y00276.1 | - | 2 - 3 | 191 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
