Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013133.1 | NP_037265.1 | 7 - 8 | 1096 | 83 | |
XM_006246378.2 | XP_006246440.1 | 7 - 8 | 1248 | 83 | ||
GenBank mRNA | AJ310834.1 | - | 7 - 8 | 912 | 83 | |
AJ310835.1 | - | 7 - 8 | 912 | 83 | ||
AJ310836.1 | - | 6 - 7 | 784 | 83 | ||
AY827463.1 | - | 7 - 8 | 912 | 83 | ||
D00833.1 | - | 7 - 8 | 1096 | 83 | ||
X55246.1 | - | 7 - 8 | 1000 | 83 | ||
Y00276.1 | - | 6 - 7 | 848 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
