Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246378.2 | XP_006246440.1 | 8 - 9 | 1397 | 89 | |
GenBank mRNA | AJ310835.1 | - | 8 - 9 | 1061 | 89 | |
X55246.1 | - | 8 - 9 | 1149 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
