Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013133.1 | NP_037265.1 | 8 - 9 | 1245 | 143 | |
GenBank mRNA | AJ310834.1 | - | 8 - 9 | 1061 | 143 | |
AJ310836.1 | - | 7 - 8 | 933 | 143 | ||
D00833.1 | - | 8 - 9 | 1245 | 143 | ||
Y00276.1 | - | 7 - 8 | 997 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
action potential
startle response
regulation of respiratory gaseous exchange by nervous system process
ion transport
anion transport
chloride transport
muscle contraction
neuropeptide signaling pathway
chemical synaptic transmission
acrosome reaction
visual perception
adult walking behavior
ion transmembrane transport
regulation of membrane potential
regulation of respiratory gaseous exchange
neuromuscular process controlling posture
neuromuscular process
protein homooligomerization
protein heterooligomerization
negative regulation of transmission of nerve impulse
synaptic transmission, glycinergic
righting reflex
inhibitory postsynaptic potential
chloride transmembrane transport
positive regulation of acrosome reaction
glycine binding
extracellularly glycine-gated ion channel activity
extracellularly glycine-gated chloride channel activity
transmitter-gated ion channel activity
taurine binding
action potential
startle response
regulation of respiratory gaseous exchange by nervous system process
ion transport
anion transport
chloride transport
muscle contraction
neuropeptide signaling pathway
chemical synaptic transmission
acrosome reaction
visual perception
adult walking behavior
ion transmembrane transport
regulation of membrane potential
regulation of respiratory gaseous exchange
neuromuscular process controlling posture
neuromuscular process
protein homooligomerization
protein heterooligomerization
negative regulation of transmission of nerve impulse
synaptic transmission, glycinergic
righting reflex
inhibitory postsynaptic potential
chloride transmembrane transport
positive regulation of acrosome reaction
glycine binding
extracellularly glycine-gated ion channel activity
extracellularly glycine-gated chloride channel activity
transmitter-gated ion channel activity
taurine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56746 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053621 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053621 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053621 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P196389 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146851 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98372 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15060 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2408 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87683 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25459 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4898 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79977 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21470 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10742830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742823 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10742820 | - | - |
Rat Genome 230 2.0 Array | 1371156_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1387464_at | - | - |
Rat Genome 230 2.0 Array | 1387464_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371156_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371156_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371156_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387464_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387464_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000024890.rn.1 | TC1000002832.rn.1 | RTA TC |
Rat Expression Set 230 | 1371156_PM_a_at | - | - |
Rat Expression Set 230 | 1371156_a_at | - | - |
Rat Expression Set 230 | 1387464_PM_at | - | - |
Rat Expression Set 230 | 1387464_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742820 | - | - |
Rat Genome U34 Set | X55246_at | - | - |
Rat Genome U34 Set | D00833_at | - | - |
Rat Genome U34 Set | D00833_g_at | - | - |
Rat Genome U34 Set | X55246_at | - | - |
Rat Genome U34 Set | D00833_g_at | - | - |
Rat Genome U34 Set | D00833_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22004636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362530 | - | - |
RatRef-12 Expression BeadChip | ILMN_56361 | - | - |
