Assay Details
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013115.2 | NP_001013133.2 | 3 - 4 | 214 | 80 | |
XM_006246477.2 | XP_006246539.1 | 4 - 5 | 351 | 80 | ||
XM_006246478.2 | XP_006246540.1 | 3 - 4 | 316 | 80 | ||
XM_006246479.2 | XP_006246541.1 | 2 - 3 | 159 | 80 | ||
XM_006246483.1 | XP_006246545.1 | 4 - 5 | 266 | 80 | ||
XM_008767812.1 | XP_008766034.1 | 4 - 5 | 400 | 80 | ||
XM_008767813.1 | XP_008766035.1 | 3 - 4 | 975 | 80 | ||
XM_008767814.1 | XP_008766036.1 | 3 - 4 | 224 | 80 | ||
XM_008767815.1 | XP_008766037.1 | 3 - 4 | 971 | 80 | ||
GenBank mRNA | AF354443.1 | - | 3 - 4 | 165 | 80 | |
FM065046.1 | - | 3 - 4 | 214 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
