Assay Details
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013115.2 | NP_001013133.2 | 4 - 5 | 319 | 91 | |
XM_006246477.2 | XP_006246539.1 | 5 - 6 | 456 | 91 | ||
XM_006246479.2 | XP_006246541.1 | 3 - 4 | 264 | 91 | ||
XM_006246483.1 | XP_006246545.1 | 5 - 6 | 371 | 91 | ||
GenBank mRNA | AF354443.1 | - | 4 - 5 | 270 | 91 | |
FM065046.1 | - | 4 - 5 | 319 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
