Assay Details
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246057.2 | XP_006246119.1 | 5 - 6 | 780 | 91 | |
XM_008767609.1 | XP_008765831.1 | 5 - 6 | 780 | 91 | ||
XM_008767610.1 | XP_008765832.1 | 5 - 6 | 690 | 91 | ||
XM_008767611.1 | XP_008765833.1 | 5 - 6 | 631 | 91 | ||
GenBank mRNA | FQ211597.1 | - | 5 - 6 | 747 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
