Assay Details
Gene Symbol: Fgf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022223.2 | NP_071559.2 | 2 - 3 | 304 | 87 | |
GenBank mRNA | AB008908.1 | - | 319 | 87 | ||
AB079500.1 | - | 2 - 3 | 304 | 87 | ||
AF348523.1 | - | 198 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
