Assay Details
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108387.2 | NP_001101857.2 | 3 - 4 | 370 | 88 | |
GenBank mRNA | BC166998.1 | - | 2 - 3 | 290 | 88 | |
BF408124.1 | - | 3 - 4 | 334 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
