Assay Details
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108387.2 | NP_001101857.2 | 8 - 9 | 1110 | 126 | |
GenBank mRNA | BC166998.1 | - | 7 - 8 | 1030 | 126 | |
CF110427.1 | - | 4 - 5 | 536 | 126 | ||
FQ216292.1 | - | 4 - 5 | 476 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
