Assay Details
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108387.2 | NP_001101857.2 | 9 - 10 | 1233 | 102 | |
GenBank mRNA | BC166998.1 | - | 8 - 9 | 1153 | 102 | |
CF110427.1 | - | 5 - 6 | 659 | 102 | ||
FQ216292.1 | - | 5 - 6 | 599 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P417104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142197 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P417104 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P417104 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P417104 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P417104 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P108982 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97063 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32104 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10781460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781453 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781452 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10781451 | - | - |
Rat Genome 230 2.0 Array | 1383438_at | - | - |
Rat Genome 230 2.0 Array | 1388975_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388975_at | - | - |
Rat Genome 230 2.0 Array | 1383438_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381671_at | - | - |
Rat Genome 230 2.0 Array | 1381671_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388975_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381671_at | - | - |
Affymetrix HT Rat Focus Array | 1383438_at | - | - |
Affymetrix HT Rat Focus Array | 1381671_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388975_at | - | - |
Affymetrix HT Rat Focus Array | 1383438_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500005676.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005677.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005679.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005680.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005675.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005672.rn.1 | TC1500000824.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005674.rn.1 | TC1500000824.rn.1 | RTA TC |
Rat Expression Set 230 | 1388975_at | - | - |
Rat Expression Set 230 | 1388975_PM_at | - | - |
Rat Expression Set 230 | 1383438_PM_at | - | - |
Rat Expression Set 230 | 1383438_at | - | - |
Rat Expression Set 230 | 1381671_at | - | - |
Rat Expression Set 230 | 1381671_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781451 | - | - |
Rat Genome U34 Set | rc_AA800194_at | - | - |
Rat Genome U34 Set | rc_AA923982_g_at | - | - |
Rat Genome U34 Set | rc_AA923982_at | - | - |
Rat Genome U34 Set | rc_AI169320_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22166992 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357638 | - | - |
