Assay Details
Gene Symbol: Loxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106047.2 | NP_001099517.2 | 9 - 10 | 2011 | 80 | |
GenBank mRNA | BC168900.1 | - | 9 - 10 | 2011 | 9032601 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
