Assay Details
Gene Symbol: Loxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106047.2 | NP_001099517.2 | 12 - 13 | 2379 | 105 | |
GenBank mRNA | BC168900.1 | - | 12 - 13 | 2379 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
epithelial to mesenchymal transition
endothelial cell proliferation
sprouting angiogenesis
transcription, DNA-templated
cellular protein modification process
receptor-mediated endocytosis
histone modification
protein deamination
collagen fibril organization
positive regulation of chondrocyte differentiation
endothelial cell migration
negative regulation of transcription, DNA-templated
response to copper ion
chromatin binding
transcription corepressor activity
protein-lysine 6-oxidase activity
scavenger receptor activity
copper ion binding
methylated histone binding
oligosaccharide binding
response to hypoxia
epithelial to mesenchymal transition
endothelial cell proliferation
sprouting angiogenesis
transcription, DNA-templated
cellular protein modification process
receptor-mediated endocytosis
histone modification
protein deamination
collagen fibril organization
positive regulation of chondrocyte differentiation
endothelial cell migration
negative regulation of transcription, DNA-templated
response to copper ion
chromatin binding
transcription corepressor activity
protein-lysine 6-oxidase activity
scavenger receptor activity
copper ion binding
methylated histone binding
oligosaccharide binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P105225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163681 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P105225 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P105225 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P105225 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P105225 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72411 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10781311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781315 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10781304 | - | - |
Rat Genome 230 2.0 Array | 1380474_at | - | - |
Rat Genome 230 2.0 Array | 1391450_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380474_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391450_at | - | - |
Affymetrix HT Rat Focus Array | 1391450_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380474_at | - | - |
Affymetrix HT Rat Focus Array | 1380474_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391450_at | - | - |
Rat Expression Set 230 | 1380474_PM_at | - | - |
Rat Expression Set 230 | 1391450_at | - | - |
Rat Expression Set 230 | 1380474_at | - | - |
Rat Expression Set 230 | 1391450_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781304 | - | - |
Rat Genome U34 Set | rc_AI180353_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351352 | - | - |
