Assay Details
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013043.2 | NP_037175.1 | 1 - 2 | 308 | 104 | |
GenBank mRNA | BC059146.1 | - | 1 - 2 | 319 | 104 | |
BC127547.1 | - | 1 - 2 | 300 | 104 | ||
CB771530.1 | - | 1 - 2 | 308 | 104 | ||
L25785.1 | - | 1 - 2 | 301 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
