Assay Details
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252478.2 | XP_006252540.1 | 4 - 5 | 359 | 65 | |
XM_224534.8 | XP_224534.7 | 5 - 6 | 605 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
