Assay Details
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017333.1 | NP_059029.1 | 4 - 5 | 1052 | 94 | |
XM_006252431.2 | XP_006252493.1 | 4 - 5 | 1333 | 94 | ||
GenBank mRNA | S65355.1 | - | 4 - 5 | 1052 | 94 | |
X57764.1 | - | 3 - 4 | 1001 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
