Assay Details
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024990.2 | NP_001020161.2 | 10 - 11 | 1207 | 124 | |
GenBank mRNA | BC091278.1 | - | 9 - 10 | 1267 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
